The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172303.2 | NP_001165774.1 | 10 - 11 | 2983 | 151 |
NM_001320756.1 | NP_001307685.1 | 11 - 12 | 2995 | 151 | |
NM_001320757.1 | NP_001307686.1 | 11 - 12 | 2998 | 151 | |
NM_032844.4 | NP_116233.2 | 10 - 11 | 2980 | 151 | |
NR_135469.1 | 9 - 10 | 2841 | 151 | ||
XM_011519751.2 | XP_011518053.1 | 5 - 6 | 1500 | 151 | |
XM_017016853.1 | XP_016872342.1 | 11 - 12 | 2969 | 151 | |
GenBank mRNA | AK027719.1 | - | 10 - 11 | 2441 | 151 |
AK123004.1 | - | 9 - 10 | 3744 | 151 | |
AK225319.1 | - | 10 - 11 | 2410 | 151 | |
BC009107.1 | - | 10 - 11 | 2437 | 151 |