The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152762.2 | NP_689975.2 | 5 - 6 | 1356 | 142 |
XM_011544887.2 | XP_011543189.1 | 5 - 6 | 1555 | 142 | |
XM_011544888.2 | XP_011543190.1 | 5 - 6 | 1353 | 142 | |
GenBank mRNA | AK057442.1 | - | 5 - 6 | 1356 | 142 |
BC104021.1 | - | 2 - 3 | 352 | 142 | |
BC104022.1 | - | 5 - 6 | 1356 | 142 |