Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSGA10IP
Entrez Gene ID
254187
Gene Name
testis specific 10 interacting protein
Gene Aliases
FAM161C
Chromosome Location
Chr.11: 65945445 - 65960437 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152762.2 NP_689975.2 7 - 8 1577 85
XM_011544887.2 XP_011543189.1 7 - 8 1815 85
XM_011544888.2 XP_011543190.1 7 - 8 1613 85
GenBank mRNA AK057442.1 - 7 - 8 1577 85
BC104021.1 - 4 - 5 573 85
BC104022.1 - 7 - 8 1577 85

View Full Product Details