Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
Chromosome Location
Chr.9: 113410644 - 113429684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278629.1 NP_001265558.1 2 - 3 556 85
NM_001278630.1 NP_001265559.1 2 - 3 413 85
NM_152786.2 NP_689999.1 2 - 3 629 85
XM_011518477.1 XP_011516779.1 2 - 3 556 85
XM_011518478.1 XP_011516780.1 2 - 3 556 85
GenBank mRNA AK301872.1 - 2 - 3 413 85
AK313867.1 - 2 - 3 545 85
BC026884.1 - 2 - 3 595 85
BC033037.2 - 2 - 3 514 85

View Full Product Details