Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
Chromosome Location
Chr.9: 113410644 - 113429684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278629.1 NP_001265558.1 7 - 8 1064 68
NM_001278630.1 NP_001265559.1 7 - 8 921 68
NM_152786.2 NP_689999.1 7 - 8 1137 68
XM_005251885.3 XP_005251942.1 5 - 6 443 68
XM_011518477.1 XP_011516779.1 7 - 8 1064 68
XM_011518478.1 XP_011516780.1 7 - 8 1064 68
XM_011518479.2 XP_011516781.1 5 - 6 719 68
GenBank mRNA AK301872.1 - 7 - 8 921 68
AK313867.1 - 7 - 8 1053 68
BC026884.1 - 7 - 8 1103 68
BC033037.2 - 7 - 8 1022 68

View Full Product Details