Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTRL
Entrez Gene ID
11064
Gene Name
centriolin
Gene Aliases
CEP1, CEP110, FAN, bA165P4.1
Chromosome Location
Chr.9: 121074778 - 121177610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007018.4 NP_008949.4 37 - 38 6304 81
XM_005251679.3 XP_005251736.1 39 - 40 6576 81
XM_017014226.1 XP_016869715.1 28 - 29 4805 81
GenBank mRNA AF083322.1 - 14 - 15 2752 81
AF513978.1 - 37 - 38 6273 81
AK074079.1 - 28 - 29 4677 81
BC137284.1 - 37 - 38 6297 81
BC137286.1 - 37 - 38 6296 81
BX640927.1 - 11 - 12 1292 81

View Full Product Details