Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CNTRL
- Entrez Gene ID
-
11064
- Gene Name
- centriolin
- Gene Aliases
-
CEP1, CEP110, FAN, bA165P4.1
- Chromosome Location
-
Chr.9: 121074778 - 121177610 on Build GRCh38
View Full Product Details