Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTRL
Entrez Gene ID
11064
Gene Name
centriolin
Gene Aliases
CEP1, CEP110, FAN, bA165P4.1
Chromosome Location
Chr.9: 121074778 - 121177610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007018.4 NP_008949.4 41 - 42 6987 128
XM_005251679.3 XP_005251736.1 43 - 44 7259 128
XM_006716940.2 XP_006717003.1 42 - 43 7118 128
XM_006716942.1 XP_006717005.1 33 - 34 5533 128
XM_011518166.1 XP_011516468.1 44 - 45 7304 128
XM_011518167.1 XP_011516469.1 43 - 44 7145 128
XM_011518168.1 XP_011516470.1 44 - 45 7229 128
XM_011518169.2 XP_011516471.1 43 - 44 7179 128
XM_011518170.1 XP_011516472.1 43 - 44 7127 128
XM_011518173.1 XP_011516475.1 32 - 33 5379 128
XM_017014223.1 XP_016869712.1 44 - 45 7190 128
XM_017014224.1 XP_016869713.1 42 - 43 7004 128
XM_017014225.1 XP_016869714.1 42 - 43 6990 128
XM_017014226.1 XP_016869715.1 32 - 33 5488 128
GenBank mRNA AF083322.1 - 18 - 19 3435 128
AK074079.1 - 32 - 33 5360 128
AL080231.1 - 1 - 2 160 128
BC137284.1 - 41 - 42 6980 128
BC137286.1 - 41 - 42 6979 128
BX640927.1 - 15 - 16 1975 128

View Full Product Details