The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130438.2 | NP_001123910.1 | 35 - 36 | 4743 | 61 |
NM_001195532.1 | NP_001182461.1 | 34 - 35 | 4683 | 61 | |
NM_003127.3 | NP_003118.2 | 35 - 36 | 4743 | 61 | |
XM_006717245.1 | XP_006717308.1 | 35 - 36 | 4650 | 61 | |
XM_006717246.1 | XP_006717309.1 | 35 - 36 | 4650 | 61 | |
XM_006717247.2 | XP_006717310.1 | 34 - 35 | 4590 | 61 | |
XM_006717248.2 | XP_006717311.1 | 35 - 36 | 4649 | 61 | |
XM_006717249.1 | XP_006717312.1 | 35 - 36 | 4650 | 61 | |
XM_006717250.1 | XP_006717313.1 | 35 - 36 | 4650 | 61 | |
XM_006717251.2 | XP_006717314.1 | 34 - 35 | 4677 | 61 | |
XM_006717252.2 | XP_006717315.1 | 34 - 35 | 4590 | 61 | |
XM_006717253.1 | XP_006717316.1 | 34 - 35 | 4590 | 61 | |
XM_006717254.2 | XP_006717317.1 | 35 - 36 | 4737 | 61 | |
XM_017015059.1 | XP_016870548.1 | 35 - 36 | 4728 | 61 | |
XM_017015060.1 | XP_016870549.1 | 34 - 35 | 4590 | 61 | |
XM_017015061.1 | XP_016870550.1 | 34 - 35 | 4648 | 61 | |
GenBank mRNA | AB191262.1 | - | 34 - 35 | 4648 | 61 |
AB209860.1 | - | 35 - 36 | 4705 | 61 | |
AL110273.1 | - | 14 - 15 | 1570 | 61 | |
BC053521.1 | - | 34 - 35 | 4632 | 61 | |
J05243.1 | - | 35 - 36 | 4703 | 61 | |
M18627.1 | - | 20 - 21 | 2577 | 61 | |
M24773.1 | - | 20 - 21 | 2577 | 61 | |
U83867.1 | - | 35 - 36 | 4670 | 61 |