The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130438.2 | NP_001123910.1 | 51 - 52 | 6849 | 60 |
NM_001195532.1 | NP_001182461.1 | 49 - 50 | 6774 | 60 | |
NM_003127.3 | NP_003118.2 | 50 - 51 | 6834 | 60 | |
XM_006717248.2 | XP_006717311.1 | 51 - 52 | 6755 | 60 | |
XM_006717249.1 | XP_006717312.1 | 50 - 51 | 6741 | 60 | |
XM_006717252.2 | XP_006717315.1 | 50 - 51 | 6696 | 60 | |
XM_006717253.1 | XP_006717316.1 | 49 - 50 | 6681 | 60 | |
XM_017015061.1 | XP_016870550.1 | 50 - 51 | 6754 | 60 | |
GenBank mRNA | AB191262.1 | - | 49 - 50 | 6739 | 60 |
AB209860.1 | - | 50 - 51 | 6796 | 60 | |
AL110273.1 | - | 29 - 30 | 3661 | 60 | |
BC053521.1 | - | 49 - 50 | 6723 | 60 | |
J05243.1 | - | 50 - 51 | 6794 | 60 | |
U83867.1 | - | 51 - 52 | 6776 | 60 | |
X86901.1 | - | 4 - 5 | 518 | 60 |