The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271816.1 | NP_001258745.1 | 6 - 7 | 3235 | 75 |
NM_014881.4 | NP_055696.3 | 5 - 6 | 3553 | 75 | |
XM_006718090.1 | XP_006718153.1 | 6 - 7 | 3247 | 75 | |
XM_011540429.1 | XP_011538731.1 | 6 - 7 | 3303 | 75 | |
GenBank mRNA | BC013124.2 | - | 5 - 6 | 3185 | 75 |
BC062582.1 | - | 5 - 6 | 3185 | 75 | |
D42045.1 | - | 5 - 6 | 3437 | 75 |