The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032242.3 | NP_115618.3 | 16 - 17 | 3280 | 73 |
XM_011512908.2 | XP_011511210.1 | 17 - 18 | 3382 | 73 | |
XM_011512909.1 | XP_011511211.1 | 17 - 18 | 3975 | 73 | |
GenBank mRNA | X87832.2 | - | 16 - 17 | 3215 | 73 |