The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032242.3 | NP_115618.3 | 18 - 19 | 3660 | 67 |
XM_011512908.2 | XP_011511210.1 | 19 - 20 | 3762 | 67 | |
XM_011512909.1 | XP_011511211.1 | 19 - 20 | 4355 | 67 | |
GenBank mRNA | BQ920360.1 | - | 2 - 3 | 387 | 67 |
X87832.2 | - | 18 - 19 | 3595 | 67 |