The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015172.3 | NP_055987.2 | 2 - 3 | 379 | 136 |
XM_005245016.4 | XP_005245073.1 | 2 - 3 | 396 | 136 | |
XM_006711238.3 | XP_006711301.1 | 2 - 3 | 396 | 136 | |
XM_017000760.1 | XP_016856249.1 | 2 - 3 | 396 | 136 | |
GenBank mRNA | AF488829.1 | - | 1 - 2 | 113 | 136 |
AL096857.1 | - | 1 - 2 | 170 | 136 | |
BC001260.1 | - | 2 - 3 | 350 | 136 |