The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018085.4 | NP_060555.2 | 4 - 5 | 570 | 82 |
GenBank mRNA | DA861528.1 | - | 4 - 5 | 533 | 82 |