The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018085.4 | NP_060555.2 | 8 - 9 | 952 | 130 |
GenBank mRNA | AF410465.1 | - | 8 - 9 | 902 | 130 |