The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304331.1 | NP_001291260.1 | 15 - 16 | 2334 | 61 |
NM_001304332.1 | NP_001291261.1 | 15 - 16 | 2334 | 61 | |
XM_006711586.2 | XP_006711649.1 | 15 - 16 | 2364 | 61 | |
XM_011510070.2 | XP_011508372.1 | 3 - 4 | 393 | 61 | |
XM_017002606.1 | XP_016858095.1 | 15 - 16 | 2364 | 61 | |
XM_017002607.1 | XP_016858096.1 | 14 - 15 | 2229 | 61 | |
XM_017002608.1 | XP_016858097.1 | 15 - 16 | 2364 | 61 | |
XM_017002609.1 | XP_016858098.1 | 14 - 15 | 2229 | 61 | |
XM_017002610.1 | XP_016858099.1 | 15 - 16 | 2364 | 61 | |
GenBank mRNA | AK126624.1 | - | 2 - 3 | 902 | 61 |
AK295757.1 | - | 9 - 10 | 1062 | 61 | |
BC132923.1 | - | 2 - 3 | 844 | 61 | |
BC132925.1 | - | 2 - 3 | 844 | 61 | |
BC144262.1 | - | 2 - 3 | 844 | 61 |