The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165412.1 | NP_001158884.1 | 11 - 12 | 1522 | 92 |
NM_001319226.1 | NP_001306155.1 | 11 - 12 | 1332 | 92 | |
NM_003998.3 | NP_003989.2 | 11 - 12 | 1525 | 92 | |
XM_011532006.2 | XP_011530308.1 | 12 - 13 | 1292 | 92 | |
XM_011532008.2 | XP_011530310.1 | 9 - 10 | 906 | 92 | |
GenBank mRNA | AK122850.1 | - | 4 - 5 | 1654 | 92 |
AK291450.1 | - | 11 - 12 | 1186 | 92 | |
BC051765.1 | - | 11 - 12 | 1525 | 92 | |
M55643.1 | - | 11 - 12 | 1239 | 92 | |
M58603.1 | - | 11 - 12 | 1452 | 92 |