The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321568.1 | NP_001308497.1 | 19 - 20 | 2366 | 97 |
NM_001321570.1 | NP_001308499.1 | 19 - 20 | 2393 | 97 | |
NM_001321572.1 | NP_001308501.1 | 19 - 20 | 2393 | 97 | |
NM_001321575.1 | NP_001308504.1 | 18 - 19 | 2333 | 97 | |
NM_001321576.1 | NP_001308505.1 | 17 - 18 | 2288 | 97 | |
NM_001321577.1 | NP_001308506.1 | 18 - 19 | 2330 | 97 | |
NM_001321578.1 | NP_001308507.1 | 17 - 18 | 2260 | 97 | |
NM_001321581.1 | NP_001308510.1 | 19 - 20 | 2434 | 97 | |
NM_001321582.1 | NP_001308511.1 | 18 - 19 | 2324 | 97 | |
NM_001321584.1 | NP_001308513.1 | 18 - 19 | 2330 | 97 | |
NM_001321585.1 | NP_001308514.1 | 19 - 20 | 2427 | 97 | |
NM_001321586.1 | NP_001308515.1 | 19 - 20 | 2390 | 97 | |
NM_001321587.1 | NP_001308516.1 | 19 - 20 | 2366 | 97 | |
NM_001321590.1 | NP_001308519.1 | 18 - 19 | 2333 | 97 | |
NM_172128.2 | NP_742126.1 | 17 - 18 | 2291 | 97 | |
XM_017008674.1 | XP_016864163.1 | 19 - 20 | 2229 | 97 | |
GenBank mRNA | AK055642.1 | - | 17 - 18 | 1809 | 97 |