Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAB1
Entrez Gene ID
10454
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 1
Gene Aliases
3'-Tab1, MAP3K7IP1
Chromosome Location
Chr.22: 39399754 - 39437127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153497.2 NP_705717.1 10 - 11 1359 83
GenBank mRNA AF425640.1 - 10 - 11 1328 83

View Full Product Details