The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194313.2 | NP_919289.2 | 5 - 6 | 1251 | 71 |
XM_011517861.1 | XP_011516163.1 | 6 - 7 | 1472 | 71 | |
XM_011517863.2 | XP_011516165.1 | 5 - 6 | 2561 | 71 | |
XM_017014697.1 | XP_016870186.1 | 5 - 6 | 1565 | 71 | |
XM_017014698.1 | XP_016870187.1 | 4 - 5 | 1338 | 71 | |
GenBank mRNA | AK310014.1 | - | 6 - 7 | 1472 | 71 |
BF895212.1 | - | 3 - 4 | 537 | 71 | |
BP871643.1 | - | 2 - 3 | 305 | 71 |