The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000453.2 | NP_000444.1 | 11 - 12 | 1678 | 71 |
XM_011528192.2 | XP_011526494.1 | 11 - 12 | 2290 | 71 | |
XM_011528193.2 | XP_011526495.1 | 12 - 13 | 1793 | 71 | |
XM_011528194.2 | XP_011526496.1 | 11 - 12 | 1198 | 71 | |
XM_017027158.1 | XP_016882647.1 | 12 - 13 | 1758 | 71 | |
GenBank mRNA | BC105047.1 | - | 11 - 12 | 1393 | 71 |
BC105049.1 | - | 11 - 12 | 1393 | 71 | |
BX648217.1 | - | 8 - 9 | 892 | 71 | |
U66088.1 | - | 11 - 12 | 1678 | 71 |