The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000453.2 | NP_000444.1 | 12 - 13 | 1877 | 93 |
XM_011528192.2 | XP_011526494.1 | 12 - 13 | 2489 | 93 | |
XM_011528193.2 | XP_011526495.1 | 13 - 14 | 1992 | 93 | |
XM_011528194.2 | XP_011526496.1 | 12 - 13 | 1397 | 93 | |
XM_017027158.1 | XP_016882647.1 | 13 - 14 | 1957 | 93 | |
GenBank mRNA | BC105047.1 | - | 12 - 13 | 1592 | 93 |
BC105049.1 | - | 12 - 13 | 1592 | 93 | |
BX648217.1 | - | 9 - 10 | 1091 | 93 | |
D87920.1 | - | 12 - 13 | 1749 | 93 | |
U66088.1 | - | 12 - 13 | 1877 | 93 |