The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000453.2 | NP_000444.1 | 3 - 4 | 819 | 77 |
XM_011528192.2 | XP_011526494.1 | 3 - 4 | 1398 | 77 | |
XM_011528193.2 | XP_011526495.1 | 4 - 5 | 901 | 77 | |
XM_011528194.2 | XP_011526496.1 | 3 - 4 | 306 | 77 | |
XM_017027158.1 | XP_016882647.1 | 4 - 5 | 899 | 77 | |
GenBank mRNA | BC105047.1 | - | 3 - 4 | 534 | 77 |
BC105049.1 | - | 3 - 4 | 534 | 77 | |
D87920.1 | - | 3 - 4 | 691 | 77 | |
U66088.1 | - | 3 - 4 | 819 | 77 |