The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000453.2 | NP_000444.1 | 5 - 6 | 1042 | 80 |
XM_011528192.2 | XP_011526494.1 | 5 - 6 | 1621 | 80 | |
XM_011528193.2 | XP_011526495.1 | 6 - 7 | 1124 | 80 | |
XM_011528194.2 | XP_011526496.1 | 5 - 6 | 529 | 80 | |
XM_017027158.1 | XP_016882647.1 | 6 - 7 | 1122 | 80 | |
GenBank mRNA | BC105047.1 | - | 5 - 6 | 757 | 80 |
BC105049.1 | - | 5 - 6 | 757 | 80 | |
BX648217.1 | - | 2 - 3 | 256 | 80 | |
D87920.1 | - | 5 - 6 | 914 | 80 | |
U66088.1 | - | 5 - 6 | 1042 | 80 |