Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP76
Entrez Gene ID
79959
Gene Name
centrosomal protein 76
Gene Aliases
C18orf9, HsT1705
Chromosome Location
Chr.18: 12661733 - 12702777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271989.1 NP_001258918.1 10 - 11 1845 108
NM_024899.3 NP_079175.2 11 - 12 2070 108
NR_073537.1 11 - 12 2065 108
XM_005258149.4 XP_005258206.1 11 - 12 2049 108
XM_017025981.1 XP_016881470.1 11 - 12 2049 108
XM_017025982.1 XP_016881471.1 11 - 12 2130 108
XM_017025983.1 XP_016881472.1 10 - 11 1920 108
XM_017025984.1 XP_016881473.1 11 - 12 2032 108
XM_017025985.1 XP_016881474.1 11 - 12 1998 108
XM_017025986.1 XP_016881475.1 10 - 11 1807 108
XM_017025987.1 XP_016881476.1 10 - 11 1774 108
GenBank mRNA AK022604.1 - 11 - 12 1932 108
AK301964.1 - 10 - 11 1727 108
AK303117.1 - 10 - 11 1792 108
BC026307.1 - 11 - 12 1997 108
BC033026.1 - 11 - 12 1932 108

View Full Product Details