Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP76
Entrez Gene ID
79959
Gene Name
centrosomal protein 76
Gene Aliases
C18orf9, HsT1705
Chromosome Location
Chr.18: 12661733 - 12702777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271989.1 NP_001258918.1 8 - 9 1291 97
NM_024899.3 NP_079175.2 9 - 10 1516 97
NR_073537.1 9 - 10 1511 97
XM_005258149.4 XP_005258206.1 9 - 10 1495 97
XM_017025981.1 XP_016881470.1 9 - 10 1495 97
XM_017025982.1 XP_016881471.1 9 - 10 1576 97
XM_017025983.1 XP_016881472.1 8 - 9 1366 97
XM_017025984.1 XP_016881473.1 9 - 10 1478 97
XM_017025985.1 XP_016881474.1 9 - 10 1444 97
XM_017025986.1 XP_016881475.1 8 - 9 1253 97
XM_017025987.1 XP_016881476.1 8 - 9 1220 97
GenBank mRNA AK022604.1 - 9 - 10 1378 97
AK303117.1 - 8 - 9 1238 97
BC026307.1 - 9 - 10 1443 97
BC033026.1 - 9 - 10 1378 97

View Full Product Details