Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABHD8
Entrez Gene ID
79575
Gene Name
abhydrolase domain containing 8
Gene Aliases
-
Chromosome Location
Chr.19: 17292131 - 17303473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024527.4 NP_078803.4 3 - 4 1177 59
GenBank mRNA AK021805.1 - 3 - 4 1129 59
AK225140.1 - 3 - 4 1103 59
AK225340.1 - 3 - 4 1130 59
AK298251.1 - 4 - 5 1059 59
AK315366.1 - 3 - 4 1117 59
BC007895.2 - 3 - 4 1079 59
BC020173.1 - 3 - 4 979 59
BC039087.1 - 3 - 4 1022 59
EU285559.1 - 2 - 3 937 59

View Full Product Details