Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKD2L2
Entrez Gene ID
27039
Gene Name
polycystin 2 like 2, transient receptor potential cation channel
Gene Aliases
TRPP5
Chromosome Location
Chr.5: 137889420 - 137942747 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258449.1 NP_001245378.1 6 - 7 1029 159
NM_001300921.1 NP_001287850.1 6 - 7 1029 159
NM_014386.3 NP_055201.2 6 - 7 1029 159
XM_011543318.2 XP_011541620.1 5 - 6 943 159
XM_011543321.1 XP_011541623.1 6 - 7 963 159
XM_017009343.1 XP_016864832.1 6 - 7 1045 159
XM_017009344.1 XP_016864833.1 5 - 6 1153 159
XM_017009345.1 XP_016864834.1 5 - 6 823 159
GenBank mRNA AF118125.1 - 6 - 7 996 159
AF182034.1 - 6 - 7 999 159
BC044581.1 - 6 - 7 1028 159

View Full Product Details