The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005142.2 | NP_005133.2 | 2 - 3 | 302 | 81 |
XM_011544939.2 | XP_011543241.1 | 2 - 3 | 330 | 81 | |
GenBank mRNA | AK301290.1 | - | 2 - 3 | 300 | 81 |
AK314275.1 | - | 2 - 3 | 321 | 81 | |
BC037958.1 | - | 2 - 3 | 302 | 81 | |
X76562.1 | - | 2 - 3 | 301 | 81 |