The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203424.1 | NP_982248.1 | 1 - 2 | 54 | 117 |
GenBank mRNA | BC040047.1 | - | 1 - 2 | 45 | 117 |