The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079514.2 | NP_001072982.1 | 14 - 15 | 2421 | 105 |
NM_001288656.1 | NP_001275585.1 | 13 - 14 | 1855 | 105 | |
XM_005255277.3 | XP_005255334.1 | 14 - 15 | 2606 | 105 | |
XM_005255278.3 | XP_005255335.1 | 14 - 15 | 3570 | 105 | |
XM_005255279.3 | XP_005255336.1 | 13 - 14 | 2459 | 105 | |
XM_011522465.1 | XP_011520767.1 | 14 - 15 | 2606 | 105 | |
XM_011522466.2 | XP_011520768.1 | 9 - 10 | 1247 | 105 | |
XM_017023189.1 | XP_016878678.1 | 14 - 15 | 2606 | 105 | |
XM_017023190.1 | XP_016878679.1 | 13 - 14 | 2459 | 105 | |
XM_017023191.1 | XP_016878680.1 | 7 - 8 | 979 | 105 | |
GenBank mRNA | AK300131.1 | - | 13 - 14 | 1855 | 105 |
U19346.1 | - | 8 - 9 | 761 | 105 |