The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079514.2 | NP_001072982.1 | 4 - 5 | 1048 | 109 |
NM_001288656.1 | NP_001275585.1 | 3 - 4 | 482 | 109 | |
XM_005255277.3 | XP_005255334.1 | 4 - 5 | 1233 | 109 | |
XM_005255278.3 | XP_005255335.1 | 4 - 5 | 2197 | 109 | |
XM_005255279.3 | XP_005255336.1 | 4 - 5 | 1233 | 109 | |
XM_011522465.1 | XP_011520767.1 | 4 - 5 | 1233 | 109 | |
XM_011522467.1 | XP_011520769.1 | 4 - 5 | 1233 | 109 | |
XM_017023189.1 | XP_016878678.1 | 4 - 5 | 1233 | 109 | |
XM_017023190.1 | XP_016878679.1 | 4 - 5 | 1233 | 109 | |
XM_017023192.1 | XP_016878681.1 | 4 - 5 | 1233 | 109 | |
GenBank mRNA | AF108460.1 | - | 4 - 5 | 543 | 109 |
AF108461.1 | - | 3 - 4 | 1130 | 109 | |
AK056282.1 | - | 4 - 5 | 2197 | 109 | |
AK300131.1 | - | 3 - 4 | 482 | 109 | |
BC013750.1 | - | 4 - 5 | 768 | 109 |