Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AREG
Entrez Gene ID
374
Gene Name
amphiregulin
Gene Aliases
AR, AREGB, CRDGF, SDGF
Chromosome Location
Chr.4: 74445098 - 74455009 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001657.3 NP_001648.1 1 - 2 309 71
GenBank mRNA BC009799.1 - 1 - 2 275 71
BC146953.1 - 1 - 2 116 71
BC146967.1 - 1 - 2 116 71
BT019866.1 - 1 - 2 61 71
HY047499.1 - 1 - 2 310 71
M30704.1 - 1 - 2 270 71

View Full Product Details