The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001657.3 | NP_001648.1 | 3 - 4 | 764 | 66 |
GenBank mRNA | BC009799.1 | - | 3 - 4 | 730 | 66 |
BC146953.1 | - | 3 - 4 | 571 | 66 | |
BC146967.1 | - | 3 - 4 | 571 | 66 | |
BT019866.1 | - | 3 - 4 | 516 | 66 | |
M30704.1 | - | 3 - 4 | 725 | 66 |