The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001869.2 | NP_001860.2 | 2 - 3 | 172 | 60 |
GenBank mRNA | AK293378.1 | - | 2 - 3 | 168 | 60 |
BC007009.1 | - | 2 - 3 | 172 | 60 | |
BC014571.1 | - | 2 - 3 | 174 | 60 | |
BC015140.1 | - | 2 - 3 | 174 | 60 | |
BT007403.1 | - | 2 - 3 | 147 | 60 | |
CD613167.1 | - | 1 - 2 | 82 | 60 | |
KU177990.1 | - | 2 - 3 | 147 | 60 | |
KU177991.1 | - | 2 - 3 | 147 | 60 | |
KU177992.1 | - | 2 - 3 | 147 | 60 | |
U19977.1 | - | 2 - 3 | 151 | 60 |