The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001869.2 | NP_001860.2 | 5 - 6 | 512 | 83 |
GenBank mRNA | AK293378.1 | - | 5 - 6 | 508 | 83 |
BC007009.1 | - | 5 - 6 | 512 | 83 | |
BC014571.1 | - | 5 - 6 | 514 | 83 | |
BC015140.1 | - | 5 - 6 | 514 | 83 | |
BT007403.1 | - | 5 - 6 | 487 | 83 | |
CD613167.1 | - | 4 - 5 | 422 | 83 | |
KU177990.1 | - | 5 - 6 | 487 | 83 | |
U19977.1 | - | 5 - 6 | 491 | 83 |