The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193570.1 | NP_001180499.1 | 1 - 2 | 187 | 83 |
NM_004078.2 | NP_004069.1 | 1 - 2 | 187 | 83 | |
GenBank mRNA | AW163500.1 | - | 1 - 2 | 187 | 83 |