Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSRP1
Entrez Gene ID
1465
Gene Name
cysteine and glycine rich protein 1
Gene Aliases
CRP, CRP1, CSRP, CYRP, D1S181E, HEL-141, HEL-S-286
Chromosome Location
Chr.1: 201483530 - 201507259 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193570.1 NP_001180499.1 3 - 4 471 97
NM_001193571.1 NP_001180500.1 3 - 4 497 97
NM_001193572.1 NP_001180501.1 3 - 4 433 97
NM_004078.2 NP_004069.1 3 - 4 471 97
GenBank mRNA AB209756.1 - 2 - 3 278 97
AK131508.1 - 1 - 2 2725 97
AK293053.1 - 3 - 4 433 97
AK302237.1 - 3 - 4 335 97
AK304415.1 - 3 - 4 335 97
AW163500.1 - 3 - 4 471 97
BC004265.1 - 2 - 3 207 97
BC007492.2 - 3 - 4 308 97
BC018806.1 - 3 - 4 326 97
BC032493.1 - 3 - 4 325 97
BT019520.1 - 2 - 3 280 97
EU668320.1 - 3 - 4 334 97
GQ472204.1 - 2 - 3 207 97
M33146.1 - 3 - 4 315 97

View Full Product Details