Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNNM4
Entrez Gene ID
26504
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Gene Aliases
ACDP4
Chromosome Location
Chr.2: 96760902 - 96811891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020184.3 NP_064569.3 2 - 3 1642 94
XM_005263914.3 XP_005263971.1 2 - 3 1559 94
XM_005263915.3 XP_005263972.1 2 - 3 1559 94
XM_011510955.2 XP_011509257.1 2 - 3 1559 94
XM_017003799.1 XP_016859288.1 2 - 3 218 94
GenBank mRNA AB046812.1 - 2 - 3 1372 94
AF202777.1 - 2 - 3 1403 94
AK293915.1 - 2 - 3 452 94
BC063295.1 - 2 - 3 1642 94

View Full Product Details