The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001203258.1 | NP_001190187.1 | 11 - 12 | 1201 | 100 |
NM_004689.3 | NP_004680.2 | 11 - 12 | 1201 | 100 | |
XM_011537305.1 | XP_011535607.1 | 11 - 12 | 1197 | 100 | |
XM_011537306.1 | XP_011535608.1 | 11 - 12 | 1197 | 100 | |
XM_011537307.1 | XP_011535609.1 | 11 - 12 | 1197 | 100 | |
XM_011537308.2 | XP_011535610.1 | 10 - 11 | 1146 | 100 | |
XM_011537309.1 | XP_011535611.1 | 10 - 11 | 1146 | 100 | |
XM_011537310.1 | XP_011535612.1 | 10 - 11 | 1146 | 100 | |
XM_017021752.1 | XP_016877241.1 | 11 - 12 | 2370 | 100 | |
XM_017021753.1 | XP_016877242.1 | 11 - 12 | 1217 | 100 | |
XM_017021754.1 | XP_016877243.1 | 11 - 12 | 2368 | 100 | |
XM_017021755.1 | XP_016877244.1 | 10 - 11 | 2322 | 100 | |
XM_017021756.1 | XP_016877245.1 | 10 - 11 | 1127 | 100 | |
XM_017021757.1 | XP_016877246.1 | 11 - 12 | 1128 | 100 | |
XM_017021758.1 | XP_016877247.1 | 10 - 11 | 1077 | 100 | |
XM_017021759.1 | XP_016877248.1 | 8 - 9 | 798 | 100 | |
GenBank mRNA | AB209348.1 | - | 4 - 5 | 390 | 100 |
AF508978.1 | - | 11 - 12 | 1034 | 100 | |
BC006177.1 | - | 1 - 2 | 59 | 100 | |
BC142941.1 | - | 11 - 12 | 1230 | 100 | |
U35113.1 | - | 11 - 12 | 1046 | 100 |