Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLBP
Entrez Gene ID
7884
Gene Name
stem-loop binding protein
Gene Aliases
HBP
Chromosome Location
Chr.4: 1692731 - 1712741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306074.1 NP_001293003.1 1 - 2 614 73
NM_006527.3 NP_006518.1 1 - 2 614 73
GenBank mRNA AK094257.1 - 1 - 2 428 73
AK300826.1 - 1 - 2 192 73
AK315635.1 - 1 - 2 185 73
BC014908.1 - 1 - 2 183 73
BC015703.1 - 1 - 2 183 73
BT007433.1 - 1 - 2 61 73
DB027411.1 - 1 - 2 221 73
U75679.1 - 1 - 2 176 73
Z71188.1 - 1 - 2 165 73

View Full Product Details