The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018990.3 | NP_061863.1 | 5 - 6 | 768 | 76 |
GenBank mRNA | AK292023.1 | - | 5 - 6 | 716 | 76 |
AK314869.1 | - | 5 - 6 | 735 | 76 | |
AL049683.1 | - | 5 - 6 | 693 | 76 | |
BC051881.1 | - | 5 - 6 | 700 | 76 |