Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAM2
Entrez Gene ID
9697
Gene Name
translocation associated membrane protein 2
Gene Aliases
-
Chromosome Location
Chr.6: 52497402 - 52577064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012288.3 NP_036420.1 5 - 6 613 76
XM_011515005.2 XP_011513307.1 5 - 6 488 76
GenBank mRNA AB621814.1 - 5 - 6 609 76
AK291751.1 - 5 - 6 499 76
AK311560.1 - 5 - 6 482 76
BC028121.1 - 5 - 6 613 76
D31762.1 - 5 - 6 539 76
KU178488.1 - 5 - 6 464 76
KU178489.1 - 4 - 5 400 76

View Full Product Details