Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAM2
Entrez Gene ID
9697
Gene Name
translocation associated membrane protein 2
Gene Aliases
-
Chromosome Location
Chr.6: 52497402 - 52577064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012288.3 NP_036420.1 6 - 7 698 82
XM_011515005.2 XP_011513307.1 6 - 7 573 82
GenBank mRNA AK291751.1 - 6 - 7 584 82
AK311560.1 - 6 - 7 567 82
BC028121.1 - 6 - 7 698 82
D31762.1 - 6 - 7 624 82
KU178488.1 - 6 - 7 549 82
KU178489.1 - 5 - 6 485 82

View Full Product Details