The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002664.2 | NP_002655.2 | 3 - 4 | 543 | 130 |
GenBank mRNA | AB208967.1 | - | 3 - 4 | 437 | 130 |
AK313756.1 | - | 3 - 4 | 465 | 130 | |
BC018549.1 | - | 3 - 4 | 449 | 130 | |
BM477542.1 | - | 3 - 4 | 560 | 130 | |
CR542056.1 | - | 3 - 4 | 381 | 130 | |
CR542079.1 | - | 3 - 4 | 381 | 130 | |
X07743.1 | - | 3 - 4 | 441 | 130 |