The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003320.4 | NP_003311.2 | 11 - 12 | 1623 | 76 |
NM_177972.2 | NP_813977.1 | 10 - 11 | 1366 | 76 | |
XM_005253109.3 | XP_005253166.1 | 10 - 11 | 1358 | 76 | |
XM_011520344.2 | XP_011518646.1 | 10 - 11 | 1413 | 76 | |
GenBank mRNA | AK293528.1 | - | 10 - 11 | 1294 | 76 |
BC075031.2 | - | 11 - 12 | 1415 | 76 | |
BC075032.2 | - | 11 - 12 | 1415 | 76 | |
U54644.1 | - | 10 - 11 | 1369 | 76 | |
U82467.1 | - | 11 - 12 | 1623 | 76 |