The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003320.4 | NP_003311.2 | 3 - 4 | 500 | 77 |
NM_177972.2 | NP_813977.1 | 2 - 3 | 243 | 77 | |
XM_005253109.3 | XP_005253166.1 | 2 - 3 | 235 | 77 | |
XM_011520344.2 | XP_011518646.1 | 2 - 3 | 290 | 77 | |
GenBank mRNA | AK293528.1 | - | 2 - 3 | 171 | 77 |
BC075031.2 | - | 3 - 4 | 292 | 77 | |
BC075032.2 | - | 3 - 4 | 292 | 77 | |
U54644.1 | - | 2 - 3 | 246 | 77 | |
U82467.1 | - | 3 - 4 | 500 | 77 |