The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006252.3 | NP_006243.2 | 7 - 8 | 1356 | 110 |
XM_017001692.1 | XP_016857181.1 | 7 - 8 | 2821 | 110 | |
XM_017001693.1 | XP_016857182.1 | 7 - 8 | 1441 | 110 | |
XM_017001694.1 | XP_016857183.1 | 8 - 9 | 1349 | 110 | |
GenBank mRNA | AI732856.1 | - | 1 - 2 | 260 | 110 |
AK314011.1 | - | 7 - 8 | 1356 | 110 | |
BC069680.1 | - | 7 - 8 | 1328 | 110 | |
BC069740.1 | - | 7 - 8 | 1329 | 110 | |
BC069823.1 | - | 7 - 8 | 1329 | 110 | |
EF056019.1 | - | 7 - 8 | 1285 | 110 | |
FJ200485.1 | - | 5 - 6 | 1049 | 110 | |
FJ200486.1 | - | 6 - 7 | 1191 | 110 | |
HQ628625.1 | - | 6 - 7 | 1143 | 110 | |
U06454.1 | - | 7 - 8 | 1349 | 110 |