The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017552.3 | NP_060022.2 | 21 - 22 | 3314 | 139 |
XM_005264372.3 | XP_005264429.2 | 22 - 23 | 3356 | 139 | |
XM_006712030.3 | XP_006712093.1 | 22 - 23 | 3356 | 139 | |
XM_006712031.3 | XP_006712094.1 | 22 - 23 | 3356 | 139 | |
XM_011532918.2 | XP_011531220.1 | 22 - 23 | 3356 | 139 | |
XM_011532920.2 | XP_011531222.1 | 21 - 22 | 3314 | 139 | |
XM_011532924.2 | XP_011531226.1 | 22 - 23 | 3356 | 139 | |
XM_011532925.2 | XP_011531227.1 | 22 - 23 | 3356 | 139 | |
GenBank mRNA | AK125718.1 | - | 12 - 13 | 2115 | 139 |
AK131301.1 | - | 9 - 10 | 1454 | 139 |